Anti-RNASEL Mouse Monoclonal Antibody [clone: 2E9]

Supplier: Genetex
GTX30638
89264-892EA 600.1 USD
89264-892
Anti-RNASEL Mouse Monoclonal Antibody [clone: 2E9]
Antibodies
Mouse monoclonal antibody to RNase L

By Western blot, this antibody recognizes a band at ~83 kDa, representing RNaseL protein. (A low-molecular mass, 37 kDa, isoform of RNase L has been described in peripheral blood mononuclear cell extracts). Suggested working dilutions: *Western Blot - 1:1,000 to 1:4,000 (ECL)ELISA - against native protein*The investigator should determine the optimal working dilution for a specific application.

Type: Primary
Antigen: RNASEL
Clonality: Monoclonal
Clone: 2E9
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR