Anti-POMP Chicken Polyclonal Antibody

Supplier: Genetex
GTX82098
89263-836EA 707.89 USD
89263-836
Anti-POMP Chicken Polyclonal Antibody
Antibodies
Chicken polyclonal to HSPC036 protein

Type: Primary
Antigen: POMP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR