Anti-IL1A Rabbit Polyclonal Antibody

Supplier: Genetex
GTX74157
89263-288EA 628.69 USD
89263-288
Anti-IL1A Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to IL1 alpha

Recommended Starting Dilutions: ELISA: Max Dilution 0.5 ug/ml Functional Assays: 1.0 - 2.0 ug/ml Western Blotting: 0.1 - 0.2 ug/ml Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: IL1A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR