Anti-Ifng Rabbit Polyclonal Antibody

Supplier: Genetex
GTX74052
89263-194EA 628.49 USD
89263-194
Anti-Ifng Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to IFN gamma

Recommended Starting Dilutions: ELISA: 0.5 ug/ml - 2.0 ug/ml Functional Assays: 4.0ng/ml - 6.0ng/ml Western Blotting: 0.1 ug/ml - 0.2 ug/ml Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: Ifng
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR