Adjustable Torch Mount Assembly for ELAN® 9000/DRC-e, PerkinElmer
Supplier: PerkinElmer
W1020668
89260-246EA 10133.78 USD
89260-246
Adjustable Torch Mount Assembly for ELAN® 9000/DRC-e, PerkinElmer
Lights Flashlights
This torch mount assembly is used with ELAN® 9000 and DRC-e ICP-MS series instruments.
The cassette torch mount is an innovative torch design that is fully modular, demountable, rugged, and easy to maintain. It does not include the demountable quartz torch or injector.