Torch Alignment Tool for ELAN 9000/6x00/DRCs and NexION 300/350, PerkinElmer
Supplier: PerkinElmer
WE015554
89260-244EA 352.01 USD
89260-244
Torch Alignment Tool for ELAN 9000/6x00/DRCs and NexION 300/350, PerkinElmer
Lights Flashlights
The torch alignment tool is for use with demountable quartz torches in ELAN® 9000, 6x00, DRC II, and DRC-e, and all NexION® 300 and 350 ICP-MS instruments.
This tool is used to align the torch in the load coil.