Heating Mantle and Control, Quart Can, Glas-Col®
Supplier: Glas-Col
100D CH095PL
89259-998EA 1205.36 USD
89259-998
Heating Mantle and Control, Quart Can, Glas-Col®
Heaters Heating Mantles
Mantle for quart cans with a basic control.
  • Fabric interior
  • Durable aluminum outer shell
  • Supports weight of quart can
Provides heat and support for a quart can and is used extensively in arson investigation and work done in metal cans with resins and adhesives. Comes complete with compact power control having solid state circuitry, positive on off switches and pilot lights.
Delivery information: 7-10 days after order.