VWR® Precision Scissors
Supplier: VWR International
89259-974
89259-974EA 391.68 USD
89259-974
VWR® Precision Scissors
Scissors
VWR® Precision Scissors are stainless steel with very sharp, fine points.
  • Anti-magnetic properties
  • Resistant to acids
  • Scissors feature fine points
Precision scissors are available in a range of lengths and order sizes, from individual to bulk.