VWR® Epoxy Coated Aluminum Test Tube Baskets, Round
Supplier: VWR International
89259-790
89259-790EA 53.08 USD
89259-790
VWR® Epoxy Coated Aluminum Test Tube Baskets, Round
Baskets Baskets for Test Tubes
VWR® Epoxy Coated Aluminum Baskets feature a flow-through design.
  • Flow-through design
  • Autoclavable
  • Durable under many laboratory environments
They are useful in holding laboratory ware for washing, storage, or autoclaving. These baskets hold up under many laboratory environments, withstanding broad temperature ranges (−220 to 120 °C).