VWR®, Aluminum Diamond Mesh Test Tube Baskets, Round
Supplier: VWR International
89259-786
89259-786EA 38.73 USD
89259-786
VWR®, Aluminum Diamond Mesh Test Tube Baskets, Round
Baskets Baskets for Test Tubes
VWR® Aluminum Diamond Mesh Baskets are made of heavy-gauge expanded aluminum that has been riveted for a longer life.
  • Rigid construction
  • Wire mesh allows full flow-thrugh of cleaning solution
  • Safe handling with no sharp edges
Strip bands cover sharp edges all around for safe handling. Expanded metal construction provides rigidity and full flow-thru of the cleaning solution. Round shape.