1 Liter Sterilizing Capsule Filter Bag
Supplier: ADVANCED SCIENTIFICS, INC.
MDI 0.2 sterilized filters on standard 2D bags. Port 1: 3/8in ID x 5/8in OD C-Flex 374 Tubing (12in). Pinch Clamp, 0.2um PES Membrane Capsule Filter. Port 2: 3/8in ID x 5/8in OD C-Flex 374 Tubing (36in). Pinch Clamp, MPC Male+Cap. Port 3: Injection Site.