MB22 Premier Low-Profile Blades, Epredia™
Supplier: Epredia
3050822
89238-770PK 282.42 USD
89238-770
MB22 Premier Low-Profile Blades, Epredia™
Microtomes and Microtome Assemblies Microtome Blades and Handles
Epredia™ MB22 Premier Disposable Low-Profile Microtome Blade is designed with a steeper angle for applications where extremely thin, high-quality sections are required.
  • Maximum cutting angle 22°