Shandon™ Premium Disposable Microtome Blades, Epredia™, Richard-Allan Scientific
Supplier: Epredia
1001593
89238-758PK 230.03 USD
89238-758 89238-760
Shandon™ Premium Disposable Microtome Blades, Epredia™, Richard-Allan Scientific
Microtomes and Microtome Assemblies Microtome Blades and Handles
Epredia™ Shandon™ premium high-profile disposable blades are available in non-coated and PTFE-coated styles.