SP Bel-Art Trigger Sprayer, with 53 mm Adapter, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F11620-0050
89238-034CS 407.38 USD
89238-034
SP Bel-Art Trigger Sprayer, with 53 mm Adapter, Bel-Art Products, a part of SP
Closures Spray Caps
Add the convenience of adjustable spray to Scienceware® wash bottles.
  • Polypropylene construction
  • Chemically resistant
  • Stainless steel spring-action pump resists corrosion
Trigger sprayers with adapters fit any Scienceware® bottle with an easy-to-fill, 53 mm wide closure.
The simple to install sprayer is perfect for OSHA compliant 16 oz. and 32 oz. Scienceware® non-vented, wide-mouth wash bottles. A stainless steel, spring-action pump in the trigger sprayer will not corrode with repeated exposure to 10% bleach disinfecting solution.
Ordering information: The adapter plus trigger sprayer adds approximately 14 cm (51/2") height to a wash bottle. Sprayer without adapter fits most bottles with a 28 mm opening. They offer convenient, adjustable spray dispensing of a wide range of chemicals including bleach, ethanol, machine oil, and water.