Stainless Steel Industrial Cabinets, SECURALL®
Supplier: Securall Cabinets
100-SS 101-SS 102-SS 103-SS 104-SS 105-SS 106-SS 107-SS
89237-816EA 5579.69 USD
89237-816 89237-818 89237-820 89237-822 89237-824 89237-826 89237-828 89237-830
Stainless Steel Industrial Cabinets, SECURALL®
Furniture Storage Cabinets
Made from 18 gauge steel with #4 brushed finish.
  • Stainless steel 304 is corrosion and acid resistant to last a lifetime and handles everyday use in industrial environments
  • Each shelf is easily adjustable and carries a 500 lbs. load capacity
  • Flush-mounted stainless steel locking door handle eliminates projecting obstacles
Perfect for controlled environments and clean room applications such as hospitals and laboratories. Durable, 18-gauge stainless steel cabinets offer extra strength. Includes 3-point recessed stainless steel paddle handle with two brass keys and self-latching doors with steel rods.
Certifications: SMaRT certified.
Ordering information: Unpainted.