Double Polypropylene Bucket Systems, 5 Gallon, Contec®
Supplier: Contec
2770-KIT 2769-KIT
89237-616CS 1040.86 USD
89237-616 89237-620
Double Polypropylene Bucket Systems, 5 Gallon, Contec®
Mops
The Contec® Double Polypropylene Bucket System is a fully autoclavable stainless steel rolling cart with two polypropylene five gallon (20 l) buckets (red, gray), and stainless steel drip cap. The durable, yet lightweight construction makes the bucket system easy to maneuver around cleanrooms.
  • Compact, lightweight design makes the system ideal for smaller cleanrooms
  • Ideal for flat and curved Contec® mops
  • Compatible with most disinfectants
The system is available with either standard casters or heavy duty casters. The use of the Double Bucket System allows for color-coded differentiation between cleaning and waste solutions.
The five gallon (20 l) autoclavable buckets are compatible with two types of wringers, curved or flat, depending on the mop used.
Ordering information: Wringers are sold separately.