Hi-Strength 90 Spray Adhesive, 3M Industrial
Supplier: 3M
7000023924
89235-834CS 426.24 USD
89235-834
Hi-Strength 90 Spray Adhesive, 3M Industrial
Adhesives
Spray adhesive can be used for decorative laminate bonding to tables, cabinets, and shelving, and for bonding kick plates.
  • One-minute dry time
  • High temperature resistance
  • Low misting
  • Precise spray control and variable width spray pattern
  • Clear
  • Aerosol can
Adhesive has high contact bond strength, one-minute dry time, high temperature resistance, low misting, precise spray control, and variable width spray pattern.