Adapter, Conversion, Ace-Thred, Ace Glass Incorporated
Supplier: Ace Glass
13290-44 13290-37 13290-47 13290-34 13290-45
89082-468EA 51.26 USD
89082-468 89082-474 89082-478 89077-630 89235-722
Adapter, Conversion, Ace-Thred, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
Adapters are designed for use with a connecting adapter to connect vials or flasks to rotary evaporators.
  • Ace-Thred bottom
  • Top [ST] outer joint
  • Manufactured from borosilicate glass
Adapters feature either a #15 or #25 Ace-Thred bottom and a variety of [ST] outer top joint sizes.