O-Ring Kit, Mixed O-Rings, Ace Glass Incorporated
Supplier: Ace Glass
7855-300
89235-686EA 714.79 USD
89235-686
O-Ring Kit, Mixed O-Rings, Ace Glass Incorporated
Seals Sealing Rings
This mixed-material o-ring kit is packaged in a clear, plastic box with a re-sealable lid that lets you see the o-rings inside
It opens to 18 compartments, each clearly marked with the o-ring material and size. The cardboard insert indicates the codes for each size, so you can easily re-order the o-rings individually as they are depleted. Includes nine different size o-rings in four different materials, 360 o-rings in all.
Ordering information: Includes Viton o-rings (sizes -010, -011, -013, -108, and -110), Silicone o-rings (sizes -010, -011, -013, -110 and -210), FETFE o-rings (sizes -008, -013, -110, -210, and -212), and Buna-N o-rings (sizes -011, -013, and -110).