Large Scale In-Line Distillation/Reflux Splitter, Ace Glass Incorporated
Supplier: Ace Glass
6087-10
89235-646EA 4764.36 USD
89235-646
Large Scale In-Line Distillation/Reflux Splitter, Ace Glass Incorporated
Distillation Components Distillation Sets
This is a large, reflux splitter installed between the boil-up pot or column and the condenser
A 0-20 mm PTFE stopcock valve allows take-off when closed and reflux return when open. The take-off terminus is a 35/25 O-Ring ball joint. The overall height is 350 mm. Standard taper 45/50 top and bottom joints. All O-Rings are Kalrez for maximum chemical resistance. Fabricated from borosilicate glass.