Ace-Thred Filter Adapter with Side Outlet, PTFE, Ace Glass Incorporated
Supplier: Ace Glass
5861-03 5861-07
89235-616EA 517.66 USD
89235-616 89235-618
Ace-Thred Filter Adapter with Side Outlet, PTFE, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This bottom adapter is designed for use with reactor bodies and a PTFE reactor.
Fabricated from PTFE, with 3/8" NPT side discharge port. Adapters have a CAPFE O-Ring to seal adapter in Ace-Thred. The #50 size is supplied with a 100 µm polyethylene support disc. It can also be replaced with a screen support using a retainer ring.
Delivery information: The #80 size is supplied with dual filter supports; a 100 µm polyethylene filter and/or a perforated support plate with retainer ring for use with a 350 µm polypropylene screen.