Reusable Steel Knives, Epredia™
Supplier: Epredia
152020
89235-244EA 354.22 USD
89235-244 89235-246 89235-250 89235-252 89235-254 89235-256 89235-258 89235-260 89235-262 89235-270 89235-274
Reusable Steel Knives, Epredia™
Knives
Use Epredia™ Reusable Steel Knives with microtomes or cryostats when the technical application dictates that requirement.
C-profile knives are used for standard paraffin and cryo techniques.
Appropriate knife case(s) should be ordered with the knife or knives, each case holds two knives.