Sodium acetate trihydrate USP, BP, Ph. Eur., FCC

Supplier: Macco Organiques S.R.O

51101030120
89234-924EA 2178.34 USD
89234-924
Sodium acetate trihydrate USP, BP, Ph. Eur., FCC
Sodium acetate trihydrate

For excipient use only.


Certifications: ISO 9001:2008
ISO 22000:2006
Kosher including Passover days
HALAL


Ordering information: Minimum order quantities may apply; contact your VWR representative for more information.

Formula: H₃CCOONa·3H₂O
Boiling Pt: 120…123 °C (1013 hPa)
Melting Pt: 58 °C
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00071557
CAS Number: 6131-90-4
EINECS: 204-823-8
Merck Index: 13,08642
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=313c1d45673fab192565649f8cbb7ddff555de39&vid=1ad37a966605626c95117735c83b16eaea381fbc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR