Calcium chloride dihydrate 97.0-103.0% USP, Ph. Eur., BP, JP, FCC

Supplier: Macco Organiques S.R.O
Warning

Synonyms: Calcium Chloride Dihydrate, Calcium dichloride

52003020120
89234-902EA 2552 USD
89234-902
Calcium chloride dihydrate 97.0-103.0% USP, Ph. Eur., BP, JP, FCC
Calcium chloride dihydrate

For excipient use only and can be used for production dialysis solutions.


This calcium chloride dihydrate is in conformity with the last edition of:
- European Pharmacopoeia
- US Pharmacopoeia
- Japanese Pharmacopoeia
- British Pharmacopoeia
- Food Chemical Codex
- Commission regulation (EC) No 629-2008


No material of animal or human origin is used in the raw materials, in the production and distribution process. Product is BSE/TSE, GMO, allergen free, suitable for vegan/vegetarian diet.


Certifications: Certificates: GMP according to ICH Q7A, HACCP, ISO 9001:2000, CEP, Kosher including Passover days, HALAL.


Ordering information: Minimum order quantities may apply; contact your VWR representative for more information.

Formula: CaCl₂·2H₂O
MW: 147.01 g/mol
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 176 °C
Density: 1.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149613
CAS Number: 10035-04-8
EINECS: 233-140-8
Merck Index: 13,01660

Order Now

Specification Test Results

Identification (PhEur/BP/USP/FCC/JP) Corresponds all required tests
Appearance (PhEur/BP) White or almost shite, crystalline powder.
Solubility (PhEur/BP) Freely soluble in water, soluble in ethanol (96%)
Assay (PhEur/BP/USP) 97.0 - 103.0%
Solution clarity (PhEur/BP/JP) Clear
Solution color (PhEur/BP/JP) Colorless
Acidity or alkalinity (PhEur/BP) Corresponds to test
Sulfates (PhEur/BP) 0.0003
Aluminum (PhEur/BP/USP) 0.0001% Max.
Barium (PhEur/BP) Corresponds to test
Iron (PhEur/BP) 0.001% Max.
Magnesium alkali metals (PhEur/BP) 0.5% Max.
Heavy metals (PhEur/BP) 0.002% Max.
Assay (USP/FCC) 99.0-107.0%
Magnesium alkali metals (USP) 1.0% Max.
Heavy Metals (USP/JP) 0.001% Max
pH (USP/JP) 4.5-9.2
Iron aluminum and phosphate(USP/JP) corresponds to teast
Organic volatile imputiries (USP) Corresponds to test
Appearance (FCC) White, hard fragments, granules, or powder.
Solubility (FCC) Soluble in water and slightly soluble in alcohol.
Magnesium alkali metals (FCC) 4.0% Max.
Arsenic (FCC) 0.003% Max.
Fluoride (FCC) 0.004% Max.
Lead (FCC) 0.0005% Max.
Appearance (JP) White odorless granules or masses.
Solubility (JP) Very soluble in water, soluble in ethanol and practically insoluble in diethyl ether.
Assay (JP) 96.7-103.3 %
Sulfates (JP) 0.024% Max
Arsenic (JP) 0.0002% Max (As2O3)
Hypochlorite (JP) Corresponds to test
Bromides (available upon request) 0.001% Max.
Particle size greater than 0.800 mm 2-5% (for information only, non-binding)
Particle size 0.800-0.355 nm 50-80% (for information only, non-binding)
Particle size less than 0.250 mm 15-48% (for information only, non-binding)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR