Contrex® CA, Acidic Liquid Detergent, Decon Labs
Supplier: Decon Labs
5701
89234-834CS 459.36 USD
89234-834
Contrex® CA, Acidic Liquid Detergent, Decon Labs
Cleaning Agents Detergents and Surface Cleansers
Contrex® CA is a concentrated acidic liquid detergent made for use in descaling, mineral removal, oxide removal or in applications where an acidic detergent is indicated
Formulated with organic acid, Contrex® CA can be used as a manual, soak or ultrasonic cleaner. It is phosphate-free and disposable straight to drain. Made with recycled packaging.