Contrex® AL, Liquid Detergent, Decon Labs
Supplier: Decon Labs
5332
89234-824CS 435.03 USD
89234-824
Contrex® AL, Liquid Detergent, Decon Labs
Cleaning Agents Detergents and Surface Cleansers
Contrex® AL is a concentrated alkaline liquid detergent formulated for use in manual, soak or ultrasonic cleaning of labware, scientific apparatus and process equipment
Contrex® AL is free-rinsing, phosphate-free and disposable straight to drain. Made with recycled packaging.