Citrad™ Citric Acid Liquid Cleaner
Supplier: Decon Labs
4401
89234-814CS 369.49 USD
89234-814
Citrad™ Citric Acid Liquid Cleaner
Cleaning Agents Detergents and Surface Cleansers
Citrad®concentrated acidic detergent is ideal for the removal of salts, scale, and metal-oxide. Also used for metal brightening and pharmaceutical-process equipment. Completely phosphate-free, free-rinsing and biodegradable
  • Completely free-rinsing
  • Biodegradable
Citrad™ concentrated acidic detergent contains a safe, organic acid and is phosphate-free, so it will not interfere with sensitive equipment or testing procedures. This concentrated formulation must be diluted with distilled, deionized, or clean potable water and can be used as a soak, scrub, or ultrasonic cleaner. Use to clean labware, stills, cleanrooms, optical parts, pharmaceutical equipment, and manufacturing equipment. Citrad™ is also effective in removing metal oxides, scale, soil, grease, chemicals, solvents, and other compounds from surfaces and equipment.