Drug Dissolution Cannulae, Cadence Science®
Supplier: Cadence Science
9880 9881
89234-240EA 1305.73 USD
89234-240 89234-242
Drug Dissolution Cannulae, Cadence Science®
Cannulae
Ideal for drug dissolution testing
Drug dissolution testing is routinely used in the pharmaceutical industry to provide critical release information for quality control purposes, in new drug development, and to predict in vivo drug release profiles. These cannulae feature 316 stainless steel hub and tubing for maximum resistance to chemical reaction.
Ordering information: Non-sterile. Packaged individually.