Deflected Point Septum Penetration Needles, Cadence Science®
Supplier: Cadence Science
4186 4188 9860 9861 9862 9863 9864 9865 9866 4170 4173 4174 4175 4176 4180 4181 4184 4185 4187 4855 4190 4191 4192 4193 4194 4195 4854 4196 4850 4851 4853
20068-586PK 611.36 USD
20068-586 20068-590 20069-010 20069-012 20069-014 20069-016 20069-018 20069-020 20069-022 89234-180 89234-182 89234-186 89234-188 89234-190 89234-192 89234-194 89234-196 89234-198 89234-200 89234-202 89234-204 89234-206 89234-208 89234-210 89234-212 89234-214 89234-216 89234-218 89234-220 89234-222 89234-224 89234-226 89234-228 89234-230
Deflected Point Septum Penetration Needles, Cadence Science®
Syringe Needles
The deflected needle features point positioning on the axis of the needle lumen
This improved point geometry provides easier penetration and minimizes coring and tracking (particularly with finer gauge needles)
Ordering information: Non-sterile. Packaged 12 needles per box.