SP Bel-Art Liquid Nitrogen Cooled Mini Mortar and Pestle Set, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H37260-0100
89233-994EA 644.86 USD
89233-994
SP Bel-Art Liquid Nitrogen Cooled Mini Mortar and Pestle Set, Bel-Art Products, a part of SP
Mortars and Pestles Mortars with Pestles
Stainless steel mortar has a welded fixture to hold a 1.5 mL microcentrifuge tube.
  • Stainless steel mortar construction
  • Nested in heavily insulated polyethylene housing
  • Tube is protected from cracking
  • Polyethylene handle included on pestle
This permits cold grinding of small amounts of tissue right in the tube resulting in reduced sample loss and collection time
Tube contents are frozen by the liquid nitrogen that surrounds the mortar. This prevents the tube from cracking by eliminating direct contact with liquid nitrogen. A 12.7 cm (5”) long stainless steel pestle with 1.9 dia. x 10.2 cmW (¾ x 4”) polyethylene handle (included) can be used to pulverize the tissue. Suction feet keep the unit stable on the workbench.
The mortar is suspended above a stainless steel cavity which holds the liquid nitrogen and is nested in a heavily insulated blue polyethylene housing.