Nunc® Low Profile 5.0 ml Universal Tubes, Thermo Scientific
Supplier: Thermo Fisher Scientific
374320 374321 374323
89233-718CS 858.69 USD
89233-718 89233-720 89233-724
Nunc® Low Profile 5.0 ml Universal Tubes, Thermo Scientific
Tubes Test Tubes
Thermo Scientific Nunc™ low profile 5.0 ml externally-threaded universal tubes provide a robust solution for solid or liquid sample storage.
  • Tubes and rack are rated for storage down to vapor phase liquid nitrogen temperatures
Available in configurations that are designed to meet most tracking and packaging requirements, the low profile 5.0 ml universal tube features 2D barcodes, graduations, a white patch for labeling, recommended working volume of 4.5 ml, and a wide mouth design for easy addition and extraction of sample material.
The externally-threaded cap design minimizes contamination risk in manual handling procedures.
Certifications: IATA. SAL 10⁻⁶.