Guardian™ Digital Airflow Monitor, Labconco®
Supplier: Labconco
9413401
89233-244EA 3455 USD
89233-244
Guardian™ Digital Airflow Monitor, Labconco®
Fume Hoods and Enclosures Chemical Fume Hoods
Guardian Digital Airflow Monitors alert the operator to low airflow conditions
Include face plate with circuit board, electrical power pack, LCD display.