VWR® Basic Apron, Yellow
Supplier: VWR International
89232-928
89232-928CS 198.7 USD
89232-928
VWR® Basic Apron, Yellow
Disposable Garments
VWR® basic aprons are designed to provide protection against light splash and provide expanded coverage versus traditional aprons.
  • Extra long (57")
  • Elastic wrists, open back, bound seams
Aprons are extra long (57") to provide additional coverage below the knee. They feature elastic wrists, open back, and bound seams for maximum protection.