JACKSON SAFETY® G60 Level 5 Cut Resistant Sleeves with Dyneema® Fiber, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Ansell Healthcare
90075
89230-500CS 1087.81 USD
89230-500
JACKSON SAFETY® G60 Level 5 Cut Resistant Sleeves with Dyneema® Fiber, KIMBERLY-CLARK PROFESSIONAL®
Workwear
For the highest level of cut protection and comfort, choose JACKSON SAFETY® G60 Level 5 Cut Resistant Sleeves with Dyneema® fiber
These sleeves are a great choice for working environments that pose the risk of cuts and abrasions. ANSI Level 3/EN388 Level 5 cut resistance. Dyneema® fiber with nylon, Lycra®fiber, and glass liner provides cut resistance and a comfortable fit. Proprietary closure on arm and wrist (patent pending). Typical uses include metal fabrication, glass handling, automotive assembly, stamping, and assembly.