Barnstead™ LabTower™ RO Water Purification Systems, Thermo Scientific
Supplier: Thermo Fisher Scientific
50132391
89230-448EA 17768.55 USD
89230-448
Barnstead™ LabTower™ RO Water Purification Systems, Thermo Scientific
Water Purification Systems Pure and Ultra Pure Water Systems
The Barnstead LabTower RO water purification systems provides up to 200L/day of Type 3 (reverse osmosis) water from tap water
The LabTower RO systems are completely free-standing units which require no bench or wall space. The unit comes standard with casters for easy moving.
The LabTower RO systems are suited to meet a wide range of applications needs. Type 3 (RO) water can be used for rinsing glassware, supplying water to autoclaves, waterbaths, and preparing or diluting buffers, reagents or media for example.
All LabTower RO systems include: quick discconnects to make changing consumables easy, built in 100L reservoir and all core consumables and tubing. LabTower RO systems are offered in two output volumes, 20L/hr or 40L/hr. Both models come standard with a 100L integrated reservoir.
Certifications: CSA, CE.
Ordering information: Units come standard with universal power supply (90-240V, 50.60Hz) and all necessary tubing to connect to water inlet supply.