Barnstead™ LabTower™ TII Water Purification Systems, Thermo Scientific
Supplier: Thermo Fisher Scientific
50132141
89230-412EA 23800 USD
89230-412
Barnstead™ LabTower™ TII Water Purification Systems, Thermo Scientific
Water Purification Systems Pure and Ultra Pure Water Systems
The Barnstead LabTower TII water purification systems provides up to 200L/day of Type 2 (10-15 megohm) water from tap water
The LabTower TII systems are completely free-standing units which require no bench or wall space. The unit comes standard with casters for easy moving.
The LabTower TII systems are suited to meet a wide range of applications needs. Type 2 water can be used for rinsing glassware, supplying water to autoclaves, waterbaths, or Type 1 systems that require pretreated feed water and preparing or diluting buffers, reagents or media for example.
All LabTower TII systems include: quick discconnects to make changing consumables easy, built in 100L reservoir and all core consumables and tubing. Standard with a 100L integrated reservoir.
Ordering information: Units come standard with universal power supply (90-240V, 50.60Hz) and all necessary tubing to connect to water inlet supply.