Barnstead™ Smart2Pure™ Water Purification Systems, Thermo Scientific
Supplier: Thermo Fisher Scientific
50129688 50129845 50129872 50129885 50129887 50129890
89230-326EA 9199.91 USD
89230-326 89230-328 89230-334 89230-340 89230-342 89230-348
Barnstead™ Smart2Pure™ Water Purification Systems, Thermo Scientific
Water Purification Systems Pure and Ultra Pure Water Systems
The Barnstead Smart2Pure water purification systems provides up to 72L/day of Type 1, ultrapure (18.2 megohm) water or Type 2 (10-15 megohm) water from tap water
All Smart2Pure systems include: quick discconnects to make changing consumables easy, and all core consumables and tubing. Smart2Pure systems are offered in three output volumes, 3L/hr, 6L/hr, and 12L/hr. The 3 and 6 LPH models come standard with an integrated reservoir. The 12 LPH model requires an external reservoir with two sizes available, 30L or 60L. The Smart2Pure systems offer highly versatile mounting options. The units can be mounted on the wall, on the bench or under the bench. For the 12 LPH model, the reservoirs may also be mounted on the wall, on the bench, or under the bench. The Smart2Pure systems are offered in a variety of configurations to meet a wide range of applications needs.
The basic Smart2Pure systems are ideal for atomic absorption, ion chromatography or other analytical procedures.
The Smart2Pure UV systems are ideal for inorganic and organic trace analysis, HPLC, or ICP-MS.
The Smart2Pure UF systems are ideal for microbiology, IVF, or monoclonal antibody applications.
The Smart2Pure UV/UF systems are ideal for molecular biology, PCR, DNA work, cell or tissue culture media.
Certifications: CSA, CE exception of 89230-328, 89230-344, 89230-346 and 89230-348.
Ordering information: Units come standard with universal power supply (90-240V, 50.60Hz) and all necessary tubing to connect to water inlet supply. The Smart2Pure 3LPH and 6LPH systems come with integrated 6L reservoir. The Smart2Pure 12LPH system requires the purchase of an external reservoir. Feed water must be tap water.