Zinc chloride, anhydrous, Reagent Grade
Supplier: VWR
|
Danger
|
Synonyms:
Zinc dichloride, Butter of zinc, Dichlorozinc, Zinc (II) chloride
N882-100G
N882-500G
89230-044CS
712.77
USD
89230-044
89230-046
Zinc chloride, anhydrous, Reagent Grade
Zinc chloride
Formula:
ZnCl₂ Boiling Pt: 732 °C (1013 hPa) Melting Pt: 283 °C Density: 2.91 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011295 CAS Number: 7646-85-7 EINECS: 231-592-0 UN: 2331 ADR: 8,III Merck Index: 13,10185 |
Specification Test Results
Ammonium (NH4) |
|
Nitrate (NO3) |
|
Sulfate (SO4) |
|
Zinc Chloride (ZnCl2) |
|
Zinc Oxide (ZnO) | 0.6 % - 0.7 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...