Cobalt(II) chloride hexahydrate 98-102% ACS

Supplier: VWR

Synonyms: Cobalt Chloride, Hexahydrate, Cobalt dichloride hexahydrate, Cobalt Chloride, 6-Hydrate, Cobaltous chloride hexahydrate

J297-100G J297-500G
89230-012CS 904.79 USD
89230-012 89230-014
Cobalt(II) chloride hexahydrate 98-102% ACS
Cobalt (II) chloride hexahydrate
Formula: CoCl₂·6H₂O
MW: 237.93 g/mol
MDL Number: MFCD00149652
CAS Number: 7791-13-1
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Assay 98% - 102%
Calcium = 0.005%
Copper = 0.002%
Insolubles = 0.01%
Iron = 0.005%
Magnesium = 0.005%
Nickel = 0.1%
Nitrate = 0.01%
Potassium = 0.01%
Sodium = 0.05%
Sulfate = 0.01%
Zinc = 0.03%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR