PURELAB® Conditioning Cartridge, ELGA LabWater
Supplier: ELGA LabWater
LC178
89221-822EA 281.65 USD
89221-822
PURELAB® Conditioning Cartridge, ELGA LabWater
Filters Filter Cartridges
The conditioning cartridge is for use in PURELAB® Option-E 5/10 water purification systems, for the conditioning stage required for reversed-phase sorbents.
  • Ensures ongoing water purification effectiveness
  • Provides essential conditioning of circulating system
The conditioning cartridge is easy to install and convenient to replace.