STS Frozen Parcel Shipper, Cold Chain Technologies
Supplier: Cold Chain Technologies
STS-U186-DIUL
89221-810EA 641.54 USD
89221-810
STS Frozen Parcel Shipper, Cold Chain Technologies
Boxes Cooling/Freezing Boxes and Bags
These parcels are designed to maintain temperature no matter the orientation of the box during transit.
  • Utilizes dry ice pellets
Containers feature superior pre-qualified temperature protection for deep freeze (below –70°C) applications lasting up to 120 hours.
Weight: 70.7 lbs
Delivery information: Includes payload and positioning boxes (for dry ice sublimation) to ensure full product protection and complete coverage of dry ice during shipping.