Pipette Controller Syringes, Hamilton Company
Supplier: Hamilton
84001
89221-468EA 529.47 USD
89221-468
Pipette Controller Syringes, Hamilton Company
Syringes Syringes for Chromatography
Hamilton Company offers the most comprehensive selection of standard and custom syringes and accessories in the industry
Pipette Controller Syringes are intended to be used in place of rubber bulbs to easily and precisely fill glass pipettes. The free-sliding plunger coarsely fills the pipette, and the knurled plunger screw completes fine volume adjustment. The Luer tip syringe termination allows easy connection of tubing to glass pipettes.
Terminations
Luer Tip (LT)- The needles are removable and fit over a ground glass hub which is tapered in the shape of a male luer. The LT termination will accept most hypodermic needles but was designed specifically for use with Hamilton Kel-F needles. This termination increases the dead volume in the syringe, which may not be appropriate for some applications. Autoclavable when disassembled.
Each is manufactured to achieve the highest level of accuracy and precision possible.
Ordering information: If you are looking for a syringe, needle point or gauge that is available but not listed here, please call Avantor to place a special order by phone.