PolarSafe™ Stainless Steel Chest Freezer Racks, Argos Technologies
Supplier: Antyila Scientific
04398-44 04398-74 04398-80 04399-45 04399-46 04399-48 04401-13 04401-14 04401-16 04401-18 04401-20 04401-23 04401-24 04401-25 04401-26 04399-56 04399-58 04398-95 04398-98 04398-47 04398-46 04398-75 04398-76 04398-77 04398-28 04398-29 04398-94 04398-97 04399-43 04399-44 04399-47 04401-31 04399-77 04399-79 04399-80 04399-81 04399-82 04401-30 04402-18 04402-22 04402-24 04402-25 04402-26
71001-400EA 160.44 USD
71001-400 71001-402 71001-404 71001-440 71001-442 71001-444 71001-446 71001-448 71001-450 71001-452 71001-454 71001-456 71001-458 71001-460 71001-462 71001-464 71001-466 71001-468 71001-470 71001-472 71001-474 71001-476 71001-478 71001-480 71001-482 89128-128 89128-130 89128-132 89128-134 89128-136 89128-138 89128-140 89128-142 89128-144 89128-150 89128-152 89128-154 89128-160 89220-494 89220-496 89220-498 89220-500 89220-502 89220-504 89220-508 89220-576 89220-578 89220-580 89220-582 89220-584 89220-586 89220-588 89220-590 89220-592 89220-594
PolarSafe™ Stainless Steel Chest Freezer Racks, Argos Technologies
Racks Cryogenic Box Racks
Argos PolarSafe™ Metal Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel
Argos PolarSafe™ Racks have been configured to fill all major ultra low (-80°) freezers on the market; racks are designed for Upright and Chest ultra low freezers.
Handles at both ends accommodate freezers with right or left handed doors. Our extensive product range will accommodate all type of sample containers including 2", 3", and 5" boxes, microtiter and deepwell plates and 15 & 50mL centrifuge tubes.
Ordering information: Additional freezer racks available. Contact your Avantor sales representative for details.