Reli® Polypropylene Non-Absorbable Sutures, MYCO Medical
Supplier: Myco Medical
P8661-M
89219-042BX 186.54 USD
89219-042 89219-046 89219-048 89219-050 89219-052 89219-054
Reli® Polypropylene Non-Absorbable Sutures, MYCO Medical
Sutures
Reli® Polypropylene Sutures feature high quality, high performance needles made from AISI 304 Stainless Steel for improved needle integrity and sharpness, smooth insertion, and patient comfort
These sutures also exhibit high knot security and tensile strength. A full range of sutures is available to meet physician preference.