KIMBLE® CaliFornia Centrifuge Tube, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
45239-100
89218-792CS 945.39 USD
89218-792
KIMBLE® CaliFornia Centrifuge Tube, DWK Life Sciences
Tubes Centrifuge Tubes
Manufactured from borosilicate glass 3.3. Tube has a permanent red stripe under the white enamel graduations for easy reading of results.
  • Scale and legend are durable white ceramic enamel
  • Top is tooled to accept snap cap 28150R-6
Conical bottom centrifuge tube is used for testing of petroleum products according to ASTM D91, D893 and D1796.
Tolerance (mL)
0 to 0.1 - ±0.02
above 0.1 to 0.3 - ±0.03
above 0.3 to 0.5 - ±0.05
above 0.5 to 1 - ±0.05
above 1 to 2 - ±0.10
above 2 to 3 - ±0.10
above 3 to 5 - ±0.20
above 5 to 10 - ±0.50
above 10 to 25 - ±1.00
above 25 - ±1.00