SP Bel-Art Sterileware Double-Ended Pestle, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F19911-0001
89218-750CS 1421.99 USD
89218-750
SP Bel-Art Sterileware Double-Ended Pestle, Bel-Art Products, a part of SP
Mortars and Pestles Pestles
Double-ended pestle has flat ends to effectively press soft tissues through screens, thereby disrupting the organ structure and producing cell isolates.
  • Double-ended design allows users to choose preferred pestle diameter
  • Chemical-resistant and autoclave-safe FDA-grade polypropylene construction
  • Individually packaged and gamma sterilized and for convenience
  • End diameters: 0.375" and 0.5"
Sterilized products are not returnable.