VWR® Heavy-Duty Vacuum Bottles with Versatile Caps
Supplier: VWR International
89217-522 89217-524 89217-526 89217-528 89217-530 89217-532
89217-522EA 68.94 USD
89217-522 89217-524 89217-526 89217-528 89217-530 89217-532
VWR® Heavy-Duty Vacuum Bottles with Versatile Caps
Bottles Plastic Bottles
Heavy-duty vacuum bottles are designed to combine ease-of-use and versatility with the highest level of strength and durability.
  • Excellent replacements for glass HPLC containers
  • Available in HDPE and polypropylene formats
  • Compatible with 50mm and 80mm versatile caps and adapters
Bottles feature thick walls that maintain full vacuum for 24 h, making them excellent replacements for glass HPLC containers, and ideal for applications involving liquid aspiration, cell harvesting, and degasification. They are compatible with 50mm and 80mm versatile caps and adapters, making it easy to customize the bottles to specific applications. Bottle material and volume are molded into the side of the bottle for easy identification, along with metric graduation marks certified accurate to ±5%. Polypropylene bottles, as well as all caps and adapters, are autoclavable at 15 PSI and 121°C for 15 min.