Digital "Immersion Type" Refractometer, PAN-1, ATAGO®
Supplier: Atago
3596
89217-504EA 3367.72 USD
89217-504
Digital "Immersion Type" Refractometer, PAN-1, ATAGO®
Refractometers Benchtop Refractometers
Model PAN-1 provide continuous and real-time readings for all types of liquids such as cleaning and metal-working fluids and fluids from food and beverage industry
  • No special equipment is necessary for Installation
  • Readings are automatically updated for every 30 seconds