Barnstead™ Smart2Pure™ Water Purification Systems
Supplier: Thermo Fisher Scientific
50129885 50129845 50129688 50129890 50129872
89217-176EA 10800 USD
89217-176 89217-188 89217-172 89217-184 89217-168
Barnstead™ Smart2Pure™ Water Purification Systems
Water Purification Systems Pure and Ultra Pure Water Systems
All-in-one, compact Barnstead™ Smart2Pure™ water systems convert tap water to both ASTM Type 1 and Type 2 water.
  • Water purification progresses through independent cartridges with Aquastop quick connects for fast replacement
  • Easy to dispense and features variable speed to control flow
  • Optional UV bulb and UF ultrafilter are available to customize ultrapure water
  • The display can be tilted for optimal reading
The Smart2Pure 3 and 6 feature a built-in 6 L reservoir to store the purified water, while the Smart2Pure 12 offers the choice of a 30 or 60 L reservoir for customizable storage capacity.
Smart integrated 6L reservoir for the Smart2Pure 3 and 6 is constructed with pigment-free materials and features a conical bottom outlet for complete draining and efficient cleaning and disinfection.
The Smart2Pure 12 reservoir options of 30 or 60 L both feature a large opening which allows easy and effective cleaning by hand. Sterile venting filter/reservoir overflow protects against contamination by microorganisms. Optional CO₂ absorber prevents TOC value increases from drawn-in CO₂. Recirculation pump protects the high purity water from bacterial growth during standstills and maintains the low conductivity value. Conical bottom outlet allows for complete draining and efficient cleaning and disinfection. Wall-mount is optional for simple space-savings.
Position system on the bench or mount on the wall.