VWR® Chest Freezer LN₂ Racks for 4⁷/₈" Boxes, Stainless Steel
Supplier: VWR International
CP-CF-6-4
89214-730EA 192.84 USD
89214-730
VWR® Chest Freezer LN₂ Racks for 4⁷/₈" Boxes, Stainless Steel
Racks Cryogenic Box Racks
Durable stainless steel freezer racks facilitate organization and maximize valuable storage space.
  • Stainless steel construction
  • Durable
  • Sturdy handles
  • Standard identification labels
Racks feature sturdy handles and come standard with identification labels for inventory purposes. Racks accommodate 4⅞" high boxes and contain locking rods. Reorder by simply scanning the QR code with your smartphone or tablet.